Learn More
Accuflow SystemsSupplier Diversity Partner TUBING SIL.1/2X11/16X3/32TUBIN DFS Item
Manufacturer:  Accuflow SystemsSupplier Diversity Partner 6041161
HELIXMARK STANDARD PLATINUM-CURED SILICONE TUBING MEETS THE NEEDS OF A BROAD RANGE OF APPLICATIONS AND IS UNIVERSALLY APPLICABLE FOR THE TRANSFER OF FLUIDS UNDER STANDARD CONDITIONS. THIS TUBING IS APPROVED IN ACCORDANCE WITH USP CLASS VI, ISO 10993-1, FDA 21CFR177-2600 AND 3.1.9 OR RP 3.1.9 WITHOUT APPEARANCE OF SOLUTION. PRODUCTION AND PACKAGING UTILIZES ISO CLASS 8 CLEANROOMS AND IS MANUFACTURED IN COMPLIANCE WITH ISO 13485 QUALITY STANDARDS. THIS TUBING IS STERILIZABLE VIA AUTOCLAVING, GAMMA RADIATION OR ETHYLENE OXIDE. EVERY PACKAGE IS DOUBLE-BAGGED AND OFF-THE-SHELF AVAILABILITY ENSURES REDUCED DELIVERY TIMES.
Catalog No. NC0142435