Learn More
Cole-Parmer Masterflex Transfer Tubing, FEP-Lined Polyethylene, 1/8" ID x 1/4" OD; 100 ft  DFS Item
Manufacturer:  Cole-Parmer 0638501
Applications: Ground water sampling and other sampling and monitoring uses that require the high purity and chemical inertness of FEP. Characteristics: Chemically inert, non-toxic FEP-liner with a durable polyethylene jacket. Rigid but bends at low temperatures. Translucent. Certification: Resin is FDA-compliant (21 CFR 177.1550). Tubing is manufactured according to GMP. Temperature range: -100 to 175°F (-73 to 80°C) Sterilize: With ethylene oxide.
Catalog No. 50-634-294