Learn More
Cole-Parmer Flexelene FX16-20N FX Flexible Polyethylene, Non-DEHP 1 x 1-1/4" Tubing, 100 Ft  DFS Item
Manufacturer:  Cole-Parmer 9615520
Applications:Food and beverage processing, instrumentation, pure-water transfer, water conditioning, Characteristics: Plasticizer-free, 78 A durometer, can be used with barbed fittings, abrasion-resistant, excellent chemical compatibility, economical. Colored Flexelene™ tubing available upon request. Certification: Produced with compounds listed by NSF International Standards 51 and 61, and are FDA compliant Temperature range:-40°F to 170°F (-40°C to 76°C) Sterilize: With ethylene oxide
Catalog No. 50-629-614