Learn More
Cole-Parmer Masterflex Transfer Tubing, High-Purity PFA-450, 1/8" ID x 3/16" OD; 25 ft  DFS Item
Manufacturer:  Cole-Parmer 0636851
Applications: Pharmaceutical, biotechnology, environmental, semiconductor processing, food and beverage, autoanalyzer, and nuclear applications. Characteristics: Superior surface smoothness and clarity, low extractables with deionized water and acids, high resistance to stress and cracking. Translucent. Resin certification: Resin is FDA compliant. Tubing is manufactured according to GMP. Temperature range: -320 to 500°F (-195 to 260°C) Sterilize: With autoclave, ethylene oxide, gamma irradiation, and using dry heat.
Catalog No. 50-634-352