Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Tubulin-Β Ab-6, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ Tubulin-Β Ab-6, Mouse Monoclonal Antibody.

Manufacturer:  Thermo Scientific™ MS1226P1

 View more versions of this product

Catalog No. MS1226P1

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Provide accurate, reproducible results with the Thermo Scientific™ Tubulin-Β Ab-6, Mouse Monoclonal Antibody.

Microtubules, the major cytoskeletal elements found in all eukaryotic cells, consist of Tubulin, which is a dimer of two 55kDa subunits: alpha and Beta. Microtubules play key roles in chromosome segregation in mitosis, intracellular transport, ciliary and flagellar bending, and structural support of the cytoskeleton.

Host Species: Mouse

Clone: TBN06; same as Tub 2.5

Isotype: IgG3

Species Reactivity: Human, Cow, Rabbit, Mouse, Chicken, Rat, Frog, Moth and Wheat. Others not known.

Epitope: Not determined

Immunogen: Purified tubulin

Molecular Weight: 55kDa

Positive Control: Ls174T, MAD109 cells; Skin or lung

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Tubulin-beta Ab-6
Monoclonal
200μg/mL
Purified Ab with BSA and Azide
Murine
IgG3
RUO
Primary
Monoclonal
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
TBN06; same as Tub 2.5
Unconjugated
Concentrated
Purified tubulin
500μL
Cytoskeletal Protein
Primary
Bovine, Chicken, Frog, Human, Moth, Murine, Rabbit, Rat, Wheat
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit