Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Tubulin-β, Rabbit Polyclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ Tubulin-β, Rabbit Polyclonal Antibody.

Specifications

Antigen Tubulin-beta
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 3
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
RB9249P0
View Documents
Thermo Scientific™
RB9249P0
Purified Ab with BSA and Azide 100μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
RB9249P1
View Documents
Thermo Scientific™
RB9249P1
Purified Ab with BSA and Azide 500μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
RB9249P
View Documents
Thermo Scientific™
RB9249P
Purified Ab with BSA and Azide 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Provide accurate, reproducible results with the Thermo Scientific™ Tubulin-β, Rabbit Polyclonal Antibody.

Microtubules, the major cytoskeletal elements found in all eukaryotic cells, consist of Tubulin, which is a dimer of two 55kDa subunits: α and Β. Microtubules play key roles in chromosome segregation in mitosis, intracellular transport, ciliary and flagellar bending, and structural support of the cytoskeleton.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Cow, Pig, Rat, Mouse, Guinea Pig, Gerbil, Frog, Chicken, Yeast, Fungi, Sea Urchin, Myxamoeba and Plasmodium.

Epitope: aa 416-430

Immunogen: A synthetic peptide derived from aa 416-430 of human tubulin-β

Molecular Weight: 55kDa

Positive Control: MAD109 cells, MCF-7 cells, skin or lung

Cellular Localization: Cytoplasmic This antibody does not cause the 10nm filaments to collapse into large lateral aggregates collecting in the cell periphery or tight juxtanuclear caps. It does not block microtubule assembly. Ab-3 does not inhibit polymerization or depolymerization of platelet tubulin in vitro. It blocks (by 70-80%) the ability of tubulin dimers (with GppNHp bound) to promote a stable inhibition of adenylyl cyclase.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation
Specifications

Specifications

Tubulin-beta
Polyclonal
Rabbit
Cytoskeletal Protein
A synthetic peptide derived from aa 416-430 of human tubulin beta
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
IVD
Primary
IgG
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit