Learn More
Neese TuffWear 275 Separates
Polyurethane coating on 200-denier nylon provides better abrasion and snag resistance
Manufacturer:  Neese 275SJORGXLG
 View more versions of this product
Catalog No. 19-067442
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Alert:
Caution!Not for use as fire protection. Avoid exposing apparel to open flame or intense heat.
Description
Description
  • Garments are cool, lightweight, durable and extremely comfortable
  • All seams are double stitched and taped (tape is heat sealed over seam)
  • All garments are safety yellow
Compliance
Meet NFPA-701 flame retardant standard.
Specifications
Specifications
Safety
X-Large
Jacket
44 to 46 in.
Orange
Nylon/Polyurethane
48 to 50 in.
121.9 to 127cm