Learn More
Neese TuffWear 275 Separates
Polyurethane coating on 200-denier nylon provides better abrasion and snag resistance
$41.50 - $52.25
Specifications
Material | Nylon, Polyurethane |
---|
Alert:
Caution!Not for use as fire protection. Avoid exposing apparel to open flame or intense heat.
Catalog Number | Mfr. No. | Product Type | Size | Color | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Size | Color | Price | Quantity & Availability | |||
19-020130
|
Neese
275AJMED |
Jacket | Medium | Yellow |
Each for $41.50
|
|
19-020131
|
Neese
275AJXLG |
Jacket | X-Large | Yellow |
Each for $41.50
Case of 25 EA for $1,037.50 |
|
19-042-250
|
Neese
275AJ5XL |
Jacket | 5X-Large | Yellow |
Each for $45.50
|
|
19-076320
|
Neese
275AJXXL |
Jacket | 2X-Large | Yellow |
Each for $41.50
|
|
19-020138
|
Neese
275CLRG |
Coat | Large | Yellow |
Each for $52.25
Case of 20 EA for $1,045.00 |
|
19-020150
|
Neese
275CXLG |
Coat | X-Large | Yellow |
Each for $52.25
|
|
19-067442
|
Neese
275SJORGXLG |
Jacket | X-Large | Orange |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
- Garments are cool, lightweight, durable and extremely comfortable
- All seams are double stitched and taped (tape is heat sealed over seam)
- All garments are safety yellow
Compliance
Meet NFPA-701 flame retardant standard.
Specifications
Nylon, Polyurethane |