Neese TuffWear 275 Separates

Polyurethane coating on 200-denier nylon provides better abrasion and snag resistance

Manufacturer: Neese Industries Inc. 275CLRG

 View more versions of this product

Catalog No. 19-020138

  null
Add to Cart

Alert

Caution!Not for use as fire protection. Avoid exposing apparel to open flame or intense heat.

  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Material Nylon/Polyurethane
    Item Description Coat
    Color Yellow
    Size Large
    Size (English) Chest 44 to 46 in.
    Size (English) Waist 40 to 42 in.
    Size (Metric) Chest 111.7 to 116.8cm
    Size (Metric) Waist 101.6 to 106.68cm

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit