Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TUG 424, Tocris™

FFA1 (GPR40) agonist

Manufacturer:  R&D Systems 3795/50

Quantity 50mg

 View more versions of this product

Catalog No. 37-955-0

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

>99%
264.32
TUG 424
C18H16O2
4-[2-(2-methylphenyl)ethynyl]-benzenepropanoic acid

Specifications

50mg
>99%
264.32
TUG 424
1082058-99-8
C18H16O2
4-[2-(2-methylphenyl)ethynyl]-benzenepropanoic acid

Safety and Handling

Recommended Storage : Store at +4°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit