Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Tug Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA517022
Description
It is not recommended to aliquot this antibody.
This gene is a candidate gene for alveolar soft part sarcoma (ASPS). It has been found that this gene is fused with transcription factor TFE3 gene in ASPS and also in renal cell carcinomas. Several alternatively spliced transcript variants of this gene have been described, but their full length nature has not been determined.
Specifications
Tug | |
Polyclonal | |
0.01M HEPES with 100 μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
Q8VBT9, Q9BZE9 | |
ASPSCR1 | |
Synthetic peptide corresponding to amino acids around residue Val542 of human Tug. | |
100 μL | |
Primary | |
Human, Mouse, Rat, Monkey | |
Antibody | |
IgG |
Immunoprecipitation, Western Blot | |
Unconjugated | |
ASPSCR1 | |
1190006K01Rik; alveolar soft part sarcoma chromosomal region candidate gene 1 protein; Alveolar soft part sarcoma chromosomal region candidate gene 1 protein homolog; alveolar soft part sarcoma chromosome region, candidate 1; alveolar soft part sarcoma chromosome region, candidate 1 (human); alveolar soft part sarcoma locus; ASPC; ASPCR1; ASPL; ASPS; Aspscr1; ASPSCR1, UBX domain containing tether for SLC2A4; RCC17; renal cell carcinoma gene on chromosome 17; renal cell carcinoma, papillary, 17; renal papillary cell carcinoma protein 17; Tether containing UBX domain for GLUT4; tether, containing a UBX domain, for GLUT4; TUG; UBX domain protein 9; UBX domain-containing protein 9; UBXD9; UBXN9 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
68938, 691026, 79058 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction