Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TULA/STS-2 Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300200AF488
Description
TULA/STS-2 Polyclonal specifically detects TULA/STS-2 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TULA/STS-2 | |
Polyclonal | |
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Cbl-interacting protein 4, gene similar to UBA containing SH3 domain10CLIP4STS-2T-cell ubiquitin ligand protein, STS2, Suppressor of T-cell receptor signaling 2, T-cell ubiquitin ligand, TULAubiquitin-associated and SH3 domain-containing protein A, ubiquitin associated and SH3 domain containing A | |
Produced in rabbits immunized with purified, recombinant TULA/STS-2 (AAH69511.1; Ala354-Asn623). | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
53347 | |
Store at 4°C in the dark. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction