Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tulobuterol

beta-2-adrenergic receptor agonist

$132.40 - $533.50

Specifications

CAS 41570-61-0
Formula Weight 227.73g/mol
Storage Requirements −30° to −10°C
Format Crystalline Powder
For Use With (Application) beta-2-adrenergic receptor agonist
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ6397503 View Documents Alfa Aesar™
J6397503
1g Each for $132.40
Order in Bulk

Request bulk or custom formats

Add to cart
 
AAJ6397506 View Documents Alfa Aesar™
J6397506
5g Each for $533.50
Order in Bulk

Request bulk or custom formats

Add to cart
 
Specifications

Specifications

41570-61-0
−30° to −10°C
beta-2-adrenergic receptor agonist
159° to 163°C
227.73g/mol
Crystalline Powder
White to Off-white
Signal Transduction Reagent
Safety and Handling

Safety and Handling

Recommended Storage : Store at -20°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit