Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TULP3 Mouse anti-Human, Clone: OTI6H4, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA504150

Catalog No. 50-167-4897


Add to cart

Description

Description

This gene encodes a member of the tubby-like protein (TULP) family. Members of this family have been identified in plants, vertebrates, and invertebrates, and they share a conserved C-terminal region of approximately 200 amino acid residues. The human and mouse TULP3 proteins share 69% amino acid sequence identity, with higher identity at the N and C termini than in the central region.
Specifications

Specifications

TULP3
Monoclonal
1.2 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
O75386
TUBL3
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
7289
Immunocytochemistry, Immunofluorescence, Western Blot
OTI6H4
Unconjugated
TULP3
Liquid
TULP3
Full length human recombit protein of human TULP3 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit