Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tumstatin/COL4A3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP30296720UL

 View more versions of this product

Catalog No. NB092411


Add to Cart

Description

Description

Tumstatin/COL4A3 Polyclonal specifically detects Tumstatin/COL4A3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Tumstatin/COL4A3
Polyclonal
PBS with 50% glycerol, pH7.3.
Rabbit
Affinity purified
RUO
1285
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Western Blot, Immunofluorescence
Unconjugated
collagen alpha-3(IV) chain, collagen IV, alpha-3 polypeptide, collagen, type IV, alpha 3 (Goodpasture antigen), Goodpasture antigen, tumstatin
A synthetic peptide corresponding to a sequence within amino acids 1600 to the C-terminus of human Tumstatin/COL4A3 (NP_000082.2).
20 μL
Primary
Human, Mouse, Rat
Purified
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit