Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tungsten foil, 0.25mm (0.01 in.) thick, 99.95% (metals basis), Alfa Aesar™

Click to view available options
Quantity:
50 x 50mm
100 x 100mm
100 x 500mm
7440-33-7
W
183.84
MFCD00011461
WFKWXMTUELFFGS-UHFFFAOYSA-N
wolfram,ion,va,tungsten, elemental,wolframium,unii-v9306cxo6g,4+ ion,tungsten, soluble compounds,tungsten, insoluble compounds,powder
23964
CHEBI:27998
tungsten
[W]
7440-33-7
W
183.84
MFCD00011461
WFKWXMTUELFFGS-UHFFFAOYSA-N
wolfram,ion,va,tungsten, elemental,wolframium,unii-v9306cxo6g,4+ ion,tungsten, soluble compounds,tungsten, insoluble compounds,powder
23964
CHEBI:27998
tungsten
[W]
Quantity 50 x 50mm
Appearance Silver Gray
Density 19.3g/cm3
Assay Percent Range 99.95% (Metals basis)
Physical Form Foil, 0.25mm (0.01 in.) thick
Percent Purity 99.95%
Chemical Name or Material Tungsten, 0.25mm (0.01 in.) thick
Assay (metals basis)

EINECSNumber : 231-143-9

TSCA : Yes

Recommended Storage : Ambient temperatures