Alfa Aesar™ Tungsten gauze, 100 mesh woven from 0.0254mm (0.001 in.) dia. wire

Manufacturer: Alfa Aesar™ 40924HB

Quantity 300x300mm

 View more versions of this product

Catalog No. AA40924HB

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-33-7
    MDL Number MFCD00011461
    Chemical Name or Material Tungsten gauze, Woven from 0.0254mm (0.001 in.) dia. wire

    Specifications

    MDL Number MFCD00011461
    Mesh Size 100 Mesh
    TSCA TSCA
    Physical Form Wire Cloth
    Quantity 300x300mm
    CAS 7440-33-7
    Chemical Name or Material Tungsten gauze, Woven from 0.0254mm (0.001 in.) dia. wire

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit