Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Tungsten powder, APS 4-6 micron, Puratronic™, 99.999% (metals basis), Mo <1ppm
Click to view available options
Quantity:
2g
10g
50g
Specifications
Quantity | 50g |
Concentration | Mo <1ppm |
UN Number | UN3089 |
Physical Form | Powder |
Percent Purity | 99.999% |
Particle Size | APS 4 to 6 Micron |
Grade | Puratronic™ |
Color | Gray |
Chemical Name or Material | Tungsten powder |
Assay | (metals basis) |
Safety and Handling
GHS H Statement
H228
Flammable solid.
P210-P240-P241-P280-P370+P378q
H228-H500
DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
EINECSNumber : 231-143-9
RTECSNumber : YO7175000
TSCA : Yes
Recommended Storage : Ambient temperatures