Alfa Aesar™ Tungsten rod, 1.5mm (0.06 in.) dia., 99.95% (metals basis)

Manufacturer: Alfa Aesar™ 42233RR

Quantity 10x10cm

 View more versions of this product

Catalog No. AA42233RR

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 7440-33-7
    MDL Number MFCD00011461
    Chemical Name or Material Tungsten rod, 1.5mm (0.06 in.) dia.

    Specifications

    CAS 7440-33-7
    TSCA TSCA
    Physical Form Rod
    MDL Number MFCD00011461
    Weight ≈3.4g/10cm
    Quantity 10x10cm
    Chemical Name or Material Tungsten rod, 1.5mm (0.06 in.) dia.
    Assay 99.95%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit