Alfa Aesar™ Tungsten rod, 4.0mm (0.16 in.) dia., Puratronic™, 99.96% (metals basis)

Manufacturer: Alfa Aesar™ 11446DM

Quantity 100mm

 View more versions of this product

Catalog No. AA11446DM

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-33-7
    MDL Number MFCD00011461
    Chemical Name or Material Tungsten rod, 4.0mm (0.16 in.) dia.
    Grade Puratronic™

    Specifications

    Physical Form Rod
    MDL Number MFCD00011461
    TSCA TSCA
    CAS 7440-33-7
    Weight ≈2.42g/cm
    Quantity 100mm
    Chemical Name or Material Tungsten rod, 4.0mm (0.16 in.) dia.
    Assay 99.96%, (metals basis)
    Grade Puratronic™

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit