Tungsten rod, 99.95% (metals basis), Alfa Aesar™

Form: approx. 6.1g/10cm

Manufacturer: Alfa Aesar 43971RR

Quantity 10 x 10cm

 View more versions of this product

Catalog No. AA43971RR

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 7440-33-7
    Physical Form Rod, 2mm (0.08 in.) dia.
    Assay Percent Range 99.95% (Metals basis)
    MDL Number MFCD00011461
    Chemical Name or Material Tungsten

    Specifications

    Quantity 10 x 10cm
    Color Silver-gray
    EINECS Number 231-143-9
    Physical Form Rod, 2mm (0.08 in.) dia.
    Assay Percent Range 99.95% (Metals basis)
    MDL Number MFCD00011461
    CAS 7440-33-7
    Appearance Silver Gray rod
    Chemical Name or Material Tungsten

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit