Alfa Aesar™ Tungsten silicide, 99.5% (metals basis)

Manufacturer: Alfa Aesar™ 1155118

Quantity 50g

 View more versions of this product

Catalog No. AA1155118

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12039-88-2
    Formula Weight 240.02
    MDL Number MFCD00049704
    Chemical Name or Material Tungsten silicide

    Specifications

    Formula Weight 240.02
    Odor Odorless
    MDL Number MFCD00049704
    TSCA TSCA
    Mesh Size −325 Mesh
    Physical Form Powder
    CAS 12039-88-2
    Melting Point 2165°C
    Quantity 50g
    Chemical Name or Material Tungsten silicide
    Assay 99.5%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit