Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Tungsten(V) dichloride triethoxide, 10% w/v in ethanol

Manufacturer:  Alfa Aesar™ 39171AC

Quantity 25mL

 View more versions of this product

Catalog No. AA39171AC


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

45000-93-9
389.94
UN2924
WCl2(OC2H5)3
MFCD00210651
Tungsten(V) dichloride triethoxide

Specifications

45000-93-9
WCl2(OC2H5)3
MFCD00210651
10% w/v in ethanol
Moisture Sensitive
25mL
Liquid
389.94
UN2924
Alcohol-like
Dark blue
Tungsten(V) dichloride triethoxide

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H314
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit