Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Tungsten(V) ethoxide 1,2-dimethoxyethane adduct, 99%

Manufacturer:  Alfa Aesar™ 4565914

Quantity 25g

 View more versions of this product

Catalog No. AA4565914


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

C10H25O5W·C4H10O2
499.26
UN1993
W(OC2H5)5·C4H10O2
MFCD16294494
Tungsten (V) ethoxide 1,2-dimethoxyethane adduct

Specifications

Liquid
W(OC2H5)5·C4H10O2
MFCD16294494
Dark yellow
25g
Tungsten (V) ethoxide 1,2-dimethoxyethane adduct
C10H25O5W·C4H10O2
499.26
UN1993
<0°C
Moisture Sensitive
99%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H360-H332-H315-H319
Highly flammable liquid and vapor.
May damage fertility or the unborn child.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P201-P309-P310
Obtain special instructions before use.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit