Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tungsten(VI) chloride, 99%, 20 mesh, ACROS Organics™

Click to view available options
Quantity:
10g
50g
100g
250g
13283-01-7
Cl6W
396.54
MFCD00011463
KPGXUAIFQMJJFB-UHFFFAOYSA-H
tungsten vi chloride,tungsten hexachloride,wolfram hexachloride,wcl6,tungsten chloride wcl6 , oc-6-11,tungsten chloride, oc-6-11,tungsten chloride 8ci,tungsten 6+ hexachloride,wolframhexachlorid,hexachloridotungsten
83301
CHEBI:37771
hexachlorotungsten
Cl[W](Cl)(Cl)(Cl)(Cl)Cl
13283-01-7
Cl6W
396.54
MFCD00011463
KPGXUAIFQMJJFB-UHFFFAOYSA-H
tungsten vi chloride,tungsten hexachloride,wolfram hexachloride,wcl6,tungsten chloride wcl6 , oc-6-11,tungsten chloride, oc-6-11,tungsten chloride 8ci,tungsten 6+ hexachloride,wolframhexachlorid,hexachloridotungsten
83301
CHEBI:37771
hexachlorotungsten
Cl[W](Cl)(Cl)(Cl)(Cl)Cl
Assay Percent Range 99%
Linear Formula WCl6
Fieser 04,569; 05,742; 06,658; 14,346; 01,61; 07,94; 08,147; 10,125
Formula Weight 396.57
Physical Form Crystalline Powder
Percent Purity ≥98.5%
Color Gray-Violet
Quantity 100g
Chemical Name or Material Tungsten(VI) chloride, 20 Mesh

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger