Tungsten(VI) Chloride, 99%, Alfa Aesar™

Manufacturer: Alfa Aesar 1233718

Quantity 50g

 View more versions of this product

Catalog No. AA1233718

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13283-01-7
    Physical Form Crystalline
    Assay Percent Range 99%
    Molecular Formula WCl6
    Formula Weight 396.57
    MDL Number MFCD00011463
    Chemical Name or Material Tungsten(VI) Chloride

    Specifications

    Formula Weight 396.57
    Quantity 50g
    DOT Information DOT Class: 8
    Health Hazard 2 Keep container in a well ventilated place. Keep away from combustible material. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.
    EINECS Number 236-293-9
    Physical Form Crystalline
    Sensitivity Moisture sensitive
    Assay Percent Range 99%
    Molecular Formula WCl6
    MDL Number MFCD00011463
    CAS 13283-01-7
    Health Hazard 1 Causes burns.
    Chemical Name or Material Tungsten(VI) Chloride

    Keep container in a well ventilated place. Keep away from combustible material. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.

    Causes burns.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit