Alfa Aesar™ Tungsten(VI) isopropoxide, 5% w/v in isopropanol, packaged under Argon in resealable ChemSeal™ bottles

Manufacturer: Alfa Aesar™ 41843AC

Quantity 25mL

 View more versions of this product

Catalog No. AA41843AC

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 52321-90-1
    Linear Formula W[OCH(CH3)2]6
    Formula Weight 538.38
    MDL Number MFCD00210652
    UN Number UN1219
    Chemical Name or Material Tungsten(VI) isopropoxide

    Specifications

    Formula Weight 538.38
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    UN Number UN1219
    Health Hazard 2 GHS H Statement
    H225-H314-H336
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.
    May cause drowsiness or dizziness.
    MDL Number MFCD00210652
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL
    Physical Form Liquid
    Packaging ChemSeal™ bottles
    Sensitivity Moisture Sensitive
    Linear Formula W[OCH(CH3)2]6
    CAS 52321-90-1
    Storage Note 1
    • Packaged under argon
    Concentration 5% w/v in isopropanol
    Quantity 25mL
    Chemical Name or Material Tungsten(VI) isopropoxide

    GHS H Statement
    H225-H314-H336
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.
    May cause drowsiness or dizziness.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Packaged under argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit