Learn More
Tungsten(VI) oxide, 99.8% (metals basis), Thermo Scientific Chemicals
Description
It is used in tungsten and tungstate manufacturing which are used as X-ray screens and for fire proofing fabrics. It is used as a ceramic pigment. Nanowires of Tungsten (VI) oxide are capable of absorbing a higher percentage of the sun radiation since it absorbs blue light.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 1473°C |
Physical Form | Powder |
Quantity | 100 g |
Assay Percent Range | 99.8% (metals basis) |
Solubility Information | Tungsten (VI) oxide is soluble in hot alkali solutions. Insoluble in water and acids. Slightly soluble in hydrofluoric acid. |
Formula Weight | 231.85 |
Density | 7.16 |
Chemical Name or Material | Tungsten(VI) oxide |
Safety and Handling
EINECSNumber | 215-231-4 |
RTECSNumber | YO7760000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.