Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tungsten(VI) oxide, 99+%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Plastic bottle
1314-35-8
O3W
231.837
MFCD00011466
ZNOKGRXACCSDPY-UHFFFAOYSA-N
tungsten trioxide,tungsten vi oxide,tungstic oxide,tungstic anhydride,tungsten oxide wo3,tungsten blue,tungstic acid anhydride,wolframic acid, anhydride,tungstentrioxide,acmc-20ajaa
14811
trioxotungsten
O=[W](=O)=O
1314-35-8
O3W
231.837
MFCD00011466
ZNOKGRXACCSDPY-UHFFFAOYSA-N
tungsten trioxide,tungsten vi oxide,tungstic oxide,tungstic anhydride,tungsten oxide wo3,tungsten blue,tungstic acid anhydride,wolframic acid, anhydride,tungstentrioxide,acmc-20ajaa
14811
trioxotungsten
O=[W](=O)=O
Assay Percent Range 99+%
Linear Formula WO3
Fieser 11,591
Merck Index 15, 9997
Formula Weight 231.85
Physical Form Powder
Percent Purity ≥99.0%
Packaging Plastic bottle
Color Green-Yellow to Yellow
Quantity 100g
Chemical Name or Material Tungsten(VI) oxide
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.