Tungsten wire, Au 99.99%, Alfa Aesar™

Form: approx. 0.005g/m (W only), 3-5 wt% Au plating

Manufacturer: Alfa Aesar 41038G1

Quantity 1m

 View more versions of this product

Catalog No. AA41038G1

  Check Availability
Add to Cart

    Chemical Identifiers

    Physical Form Wire, Gold Plated, 0.025mm (0.001 in.) dia.
    Assay Percent Range 99.99% (Metals basis)
    MDL Number MFCD00011461
    Chemical Name or Material Tungsten

    Specifications

    Physical Form Wire, Gold Plated, 0.025mm (0.001 in.) dia.
    Quantity 1m
    Color Gold colored
    Density 19.3g/cm3
    Assay Percent Range 99.99% (Metals basis)
    MDL Number MFCD00011461
    Odor Odorless
    Appearance Gold colored wire
    Chemical Name or Material Tungsten

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit