Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tungsten wire basket; # turns, 8; ID(mm), 7; Ht(mm), 9; Wire leads(cm), 5.0; Wire dia(mm), 0.51, Alfa Aesar™

Click to view available options
Quantity:
10 pc.
50 pc.
7440-33-7
W
183.84
MFCD00011461
WFKWXMTUELFFGS-UHFFFAOYSA-N
wolfram,ion,va,tungsten, elemental,wolframium,unii-v9306cxo6g,4+ ion,tungsten, soluble compounds,tungsten, insoluble compounds,powder
23964
CHEBI:27998
tungsten
[W]
7440-33-7
W
183.84
MFCD00011461
WFKWXMTUELFFGS-UHFFFAOYSA-N
wolfram,ion,va,tungsten, elemental,wolframium,unii-v9306cxo6g,4+ ion,tungsten, soluble compounds,tungsten, insoluble compounds,powder
23964
CHEBI:27998
tungsten
[W]
Quantity 50 pc.
Physical Form Wire
Chemical Name or Material Tungsten wire basket, 8 turns, ID(mm), 7; Ht(mm), 9; Wire leads(cm), 5.0; Wire dia(mm), 0.51

EINECSNumber : 231-143-9

TSCA : Yes