Alfa Aesar™ Tungsten wire basket; # turns, 8; ID(mm), 9; Ht(mm), 14; Wire leads(cm), 4.0; Wire dia(mm), 0.76

For vacuum deposition

Manufacturer: Alfa Aesar™ 41176KK

Quantity 10 pieces

 View more versions of this product

Catalog No. AA41176KK

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-33-7
    MDL Number MFCD00011461
    Chemical Name or Material Tungsten wire basket, 8 turns, ID(mm), 9; Ht(mm), 14; Wire leads(cm), 4.0; Wire dia(mm), 0.76

    Specifications

    Physical Form Wire
    MDL Number MFCD00011461
    CAS 7440-33-7
    Quantity 10 pieces
    Chemical Name or Material Tungsten wire basket, 8 turns, ID(mm), 9; Ht(mm), 14; Wire leads(cm), 4.0; Wire dia(mm), 0.76

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit