Tungstosilicic acid hydrate, Alfa Aesar™

Manufacturer: Alfa Aesar 3965118

Quantity 50g

 View more versions of this product

Catalog No. AA3965118

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12027-43-9
    Physical Form Crystalline
    Molecular Formula H4SiO4·12WO3·xH2O
    Formula Weight 2878.28 (Anhydrous)
    Synonym Silicotungstic acid
    Chemical Name or Material Tungstosilicic acid hydrate, Reagent Grade

    Specifications

    Formula Weight 2878.28 (Anhydrous)
    Quantity 50g
    Physical Form Crystalline
    Molecular Formula H4SiO4·12WO3·xH2O
    CAS 12027-43-9
    Synonym Silicotungstic acid
    Chemical Name or Material Tungstosilicic acid hydrate, Reagent Grade

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit