Learn More
NSI Environmental Solutions Turbidity QC Concentrate CRM
SDP
Manufacturer:  NSI Environmental Solutions QCI014
Encompass_Preferred
A 21 mL concentrate for determination of Turbidity in the NELAC range of 0.5 - 8 NTU. Formazin based. Each container produces 2 liters of sample. Sample derived from interlab PT program.
Catalog No. NC9385472