Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Turbidity Standard 100 NTU (2020i / TC3000i), LaMotte™

Click to view available options
Quantity:
60mL
9003-70-7,7732-18-5
C18H18
234.342
CHRJZRDFSQHIFI-UHFFFAOYSA-N
174664
1,2-bis(ethenyl)benzene;styrene
C=CC1=CC=CC=C1.C=CC1=CC=CC=C1C=C
9003-70-7,7732-18-5
C18H18
234.342
CHRJZRDFSQHIFI-UHFFFAOYSA-N
174664
1,2-bis(ethenyl)benzene;styrene
C=CC1=CC=CC=C1.C=CC1=CC=CC=C1C=C
Physical Form Liquid
Color Colorless
Quantity 60mL
Chemical Name or Material Turbidity Standard, 100 NTU (2020i / TC3000i)