Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
GSC Turbidity Tube
SCI_ED
GSC Turbidity Tube measures the cloudiness of a fluid
Supplier:  GSC TURBTUB
Catalog No. S38986
Only null left
Add to Cart
Description
Description
  • Fill the tube with water and drain it slowly by limiting flow with the pinch clamp on the hose
  • Stop draining once the black and white Secchi disk on the bottom of the tube becomes visible
  • Take note of the water level using the centimeter graduations on the tube
Specifications
Specifications
Secchi disc
10 cm
0.65 kg
Plastic
60 cm
1 Year
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Your feedback has been submitted: Thank you for helping us improve our website.