Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caframo™ Blade - Turbine 2 in. (51 mm) dia with 3/8 in. (9.5 mm) bore

Blade - Turbine 2 in. (51 mm) dia with 3/8 in. (9.5 mm) bore - with set screws 316 SS electropolished. For high shear mixing with moderate vortexing.

Manufacturer:  Caframo™ A553

Catalog No. 14-501-100


Add to cart

Description

Description

  • Turbine 3 blade propeller.
  • Electropolished 316L stainless steel.
  • High shear with moderate vortexing characteristics
Specifications

Specifications

2 in.
50 mm
Caframo™ and Fisherbrand™ Overhead Stirrers
316L Stainless Steel
0.37 in.
9.5 mm
Blade only (shaft sold separately)
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit