Learn More
Cole-Parmer Davis Instruments 271 LED Miniature Vane Anemometer  DFS Item
Manufacturer:  Cole-Parmer 0595175
This weather- and shock-resistant meter features LED display, sapphire-jewel turbine bearings and an infrared rotation sensor to reduce friction. Front-panel button switches from ft/min to m/s, mph, or knots. Operates on three 1.5 V AAA batteries (not included; order separately.)
Catalog No. 50-628-416