Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TUSC3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5104363
Description
Antibody detects endogenous levels of total TUSC3.
Made up of nearly 146 million bases, chromosome 8 encodes about 800 genes. Translocation of portions of chromosome 8 with amplifications of the c-Myc gene are found in some leukemias and lymphomas, and typically associated with a poor prognosis. Portions of chromosome 8 have been linked to schizophrenia and bipolar disorder. Trisomy 8, also known as Warkany syndrome 2, most often results in early miscarriage but is occasionally seen in a mosaic form in surviving patients who suffer to a varying degree from a number of symptoms including retarded mental and motor development, and certain facial and developmental defects. WRN is a DNA helicase encoded by chromosome 8 and shown defective in those with the early aging disorder Werner syndrome. Chromosome 8 is also associated with Pfeiffer syndrome, congenital hypothyroidism and Waardenburg syndrome.
Specifications
TUSC3 | |
Polyclonal | |
Unconjugated | |
TUSC3 | |
AU022242; BC003311; D8S1992; Dolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit TUSC3; M33; Magnesium uptake/transporter TUSC3; MGC13453; MRT22; MRT7; N33; Oligosaccharyl transferase subunit TUSC3; oligosaccharyltransferase 3 homolog A; OST3A; Protein N33; Putative prostate cancer tumor suppressor; tumor suppressor candidate 3; Tusc3 | |
Rabbit | |
Affinity chromatography | |
RUO | |
290783, 7991, 80286 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q13454, Q8BTV1 | |
TUSC3 | |
A synthesized peptide derived from human TUSC3(Accession Q13454), corresponding to amino acid residues Y133-A183. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction