TVI Corporation FlexAir™ Accessories for PAPR Systems: Cape-Style

Manufacturer: TVI CorporationSupplier Diversity Partner S40084B01

 View more versions of this product

Catalog No. 19-174-108

  Check Availability
Add to Cart

    Specifications

    For Use With (Equipment) Powered air purifying respirator system
    Product Type Loose Fitting Hood
    Includes Cape
    Material Tychem™ QC
    Size Universal
    Certifications CE, NIOSH
    Color Blue

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit