TVI Corporation Pre-Filter Assemblies
Manufacturer: TVI CorporationSupplier Diversity Partner WSFSB
 View more versions of this product
Catalog No. 19-120-2393
  Check Availability
Add to Cart
    Specifications
    Includes Filter 0.75 in. x 3 ft. hose and 0.75 in. female cam to 0.75 in. male cam
    Product Type Prefilter assembly
    Material Stainless Steel
    For Use With (Application) Connection of hospital Water source to solution mixer
  • Description & Specifications
Related Products
Product Content Feedback