TVI Corporation Pre-Filter Assemblies
Manufacturer: TVI CorporationSupplier Diversity Partner WSFSA
 View more versions of this product
Catalog No. 19-065-320
  Check Availability
Add to Cart
    Specifications
    Includes Filter 1 in. x 3 ft. hose and 1 in. female cam to 1 in. male-NST connector
    Product Type Prefilter assembly
    Material Stainless Steel
    For Use With (Application) Connection of supply Water pump to flash Water heater
    For Use With (Equipment) Supply pump connection assembly
  • Description & Specifications
Related Products
Product Content Feedback