Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TVP23C Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571088
Description
This target displays homology in the following species: Cow: 79%; Dog: 79%; Horse: 86%; Human: 100%; Rabbit: 86%; Rat: 79%.
TVP23C is a protein coding gene. Gene Ontology (GO) annotations related to this gene include integral component of membrane.
Specifications
TVP23C | |
Polyclonal | |
Unconjugated | |
TVP23C | |
FAM18B2; family with sequence similarity 18, member B2; Golgi apparatus membrane protein TVP23 homolog C; Nbla10383; protein FAM18B2; trans-golgi network vesicle protein 23 homolog C (S. cerevisiae); TVP23C | |
Rabbit | |
Affinity chromatography | |
RUO | |
201158 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96ET8 | |
TVP23C | |
synthetic peptide directed towards the N-terminal region of human TVP23C. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction