Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TWEAKR/TNFRSF12 Rabbit anti-Human, Clone: SN20-07, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP267729
Description
TWEAK R/TNFRSF12 Monoclonal specifically detects TWEAK R/TNFRSF12 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TWEAKR/TNFRSF12 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500-1:1000, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin 1:50-1:200 | |
CD266, CD266 antigen, FGF-inducible 14, Fibroblast growth factor-inducible immediate-early response protein 14, FN14tumor necrosis factor receptor superfamily member 12A, tumor necrosis factor receptor superfamily, member 12A, TweakR, Tweak-receptor, type I transmembrane protein Fn14 | |
Rabbit | |
Protein A purified | |
Angiogenesis, Apoptosis, Biologically Active Proteins, Cancer, Tumor Suppressors | |
51330 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
SN20-07 | |
Unconjugated | |
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide | |
TNFRSF12A | |
Synthetic peptide within Human TWEAK R/TNFRSF12 aa 80-129/ 129. (SwissProt: Q9NP84 Human; SwissProt: Q9CR75 Mouse; Entrez Gene: 302965 Rat) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction