Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Tween 20™, Ultrapure, Thermo Scientific™

Click to view available options
Quantity:
500mL
1 gal.
Packaging:
Plastic bottle
9005-64-5
C26H50O10
522.676
MFCD00165986
HMFKFHLTUCJZJO-UHFFFAOYSA-N
Polyoxyethylene sorbitan monolaurate
443314
2-[2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy]ethyl dodecanoate
CCCCCCCCCCCC(=O)OCCOCC(C1C(C(CO1)OCCO)OCCO)OCCO
9005-64-5
C26H50O10
522.676
MFCD00165986
HMFKFHLTUCJZJO-UHFFFAOYSA-N
Polyoxyethylene sorbitan monolaurate
443314
2-[2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy]ethyl dodecanoate
CCCCCCCCCCCC(=O)OCCOCC(C1C(C(CO1)OCCO)OCCO)OCCO
Packaging Plastic bottle
Physical Form Liquid
Grade Ultrapure
Boiling Point >100°C
Flash Point >110°C (230°F)
Quantity 1 gal.
Specific Gravity 1.105
Chemical Name or Material Tween 20™