Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polysorbate 40, Acros Organics™

Click to view available options
Quantity:
25mL
250mL
1L
2.5L
Packaging:
Glass bottle
9005-66-7
C26H50O10
522.676
MFCD00165345
HMFKFHLTUCJZJO-OQUNMALSSA-N
Polyoxyethylene (20) Sorbitan Monopalmitate,Polysorbate 40
92329579
2-[(2R)-2-[(2R,3R,4R)-3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy]ethyl dodecanoate
CCCCCCCCCCCC(=O)OCCOCC(C1C(C(CO1)OCCO)OCCO)OCCO
9005-66-7
C26H50O10
522.676
MFCD00165345
HMFKFHLTUCJZJO-OQUNMALSSA-N
Polyoxyethylene (20) Sorbitan Monopalmitate,Polysorbate 40
92329579
2-[(2R)-2-[(2R,3R,4R)-3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy]ethyl dodecanoate
CCCCCCCCCCCC(=O)OCCOCC(C1C(C(CO1)OCCO)OCCO)OCCO
Density 1.0500g/mL
Hydroxyl Value 89 to 105mg KOH/g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Viscous Liquid or Paste At 25°C
Saponification Value 41 to 52mg KOH/g
Solubility Solubility in water: 100g/L. Other solubilities: 100g/L isopropyl alcohol, xylene, insoluble in propylene glycol and mineral oil
Viscosity ca.200 mPa.s (50°C)
Color Yellow
Flash Point >200°C
Quantity 250mL
Specific Gravity 1.05
Chemical Name or Material Polysorbate 40
Show More Show Less