Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ TWEEN™ 80, Protein Grade™ Detergent, 10% Solution, Sterile-Filtered, Calbiochem™,

Catalog Number 65520750ml
Click to view available options
Quantity:
50 mL
Packaging:
Glass Bottle
9005-65-6
(C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6
604.82
MFCD00082107
HDTIFOGXOGLRCB-KTKRTIGZNA-N
Polysorbate 80
2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate
CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
9005-65-6
(C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6
604.82
MFCD00082107
HDTIFOGXOGLRCB-KTKRTIGZNA-N
Polysorbate 80
2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate
CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit