Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polysorbate 85, Acros Organics™

Click to view available options
Quantity:
25mL
250mL
1L
2.5L
Packaging:
Glass bottle
9005-70-3
(C2H4O)D(C2H4O)C(C2H4O)B(C2H4O)AC60H107O8
Polyoxyethylene (20) Sorbitan Trioleate,Polysorbate 85
9005-70-3
(C2H4O)D(C2H4O)C(C2H4O)B(C2H4O)AC60H107O8
Polyoxyethylene (20) Sorbitan Trioleate,Polysorbate 85
Acid Value 2mg KOH/g max.
Density 1.0200g/mL
Dioxane 5ppm max.
Hydroxyl Value 38 to 52mg KOH/g
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4700 to 1.4720
Physical Form Liquid
Saponification Value 83 to 98mg KOH/g
Solubility Solubility in water: soluble. Other solubilities: 100g/L isopropyl alcohol, xylene and mineral oil, insoluble in propylene glycol
Viscosity 300 mPa.s (25°C)
Water 3% Max.
Color Colorless
Flash Point >200°C
Quantity 25mL
Specific Gravity 1.02
Chemical Name or Material Polysorbate 85
Show More Show Less