Learn More
TWF1 Mouse anti-Human, Clone: OTI5C11, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF808892
Catalog No. 50-166-9156
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes twinfilin, an actin monomer-binding protein conserved from yeast to mammals. Studies of the mouse counterpart suggest that this protein may be an actin monomer-binding protein, and its localization to cortical G-actin-rich structures may be regulated by the small GTPase RAC1.
Specifications
Specifications
TWF1
Monoclonal
Unconjugated
TWF1
Lyophilized
TWF1
Human recombit protein fragment corresponding to amino acids 140-384 of human TWF1 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI5C11
PBS with 8% trehalose and no preservative; pH 7.3
Q12792
A6, PTK9
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
5756