Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ twin.tec™ Real-Time 96-Well PCR Platesc

Fluorescence intensity up to ten times higher than with frosted wells

Specifications

Autoclavable Autoclavable
Barcode Yes
For Use With (Application) RT-PCR applications
Well Color White
Material Polypropylene
View More Specs

Products 1
Catalog Number Mfr. No. Color Type Skirt Style Price Quantity & Availability  
Catalog Number Mfr. No. Color Type Skirt Style Price Quantity & Availability  
14-281-213
View Documents
Eppendorf™
0030132750
Black This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

The limiting factor in low volume real-time PCR is often the remaining intensity of fluorescence. The white wells of the Eppendorf twin.tec real-time PCR plates reflect fluorescence much better than clear or frosted wells. Thus, lower levels of fluorescence are still detectable with the same instrument–just by using the right consumables. Additionally, white wells significantly reduce interfering background fluorescence and lead to increased homogeneity of replicates and reproducible results.

  • White wells for better reflection
  • High mechanical stability
  • Raised rims for effective sealing
  • Optimal heat transfer due to reduced wall thickness
  • Autoclavable (121°C, 20 min.)
  • Rigid polycarbonate frame ensures easy and reliable handling, manually or automated
  • Polypropylene wells guarantee excellent and fast temperature transfer to the sample

Specifications

Specifications

Autoclavable
RT-PCR applications
Polypropylene
Sterile
Yes
White
Yes
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit